missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromosalicylaldehyde, 98%

Product Code. 10045853 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10045853 1 g Glass bottle 1g
10562551 25 g Glass bottle 25g
10562741 100 g Glass bottle 100g
10589243 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10045853 Supplier Thermo Scientific Acros Supplier No. 153400010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 90-59-5 | C7H4Br2O2 | 279.915 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-59-5
MDL Number MFCD00003318
InChI Key JHZOXYGFQMROFJ-UHFFFAOYSA-N
Synonym 3,5-dibromosalicylaldehyde, dalyde, 3,5-dibromosalicylaldeyde, benzaldehyde, 3,5-dibromo-2-hydroxy, 2-hydroxy-3,5-dibromobenzaldehyde, 3,5-dibromosilicylaldehyde, unii-7a260pbi3a, salicylaldehyde, 3,5-dibromo, 3,5-dibromsalicylaldehyd, 3,5-dibromo salicyclic aldehyde
PubChem CID 7024
IUPAC Name 3,5-dibromo-2-hydroxybenzaldehyde
SMILES C1=C(C=C(C(=C1Br)O)C=O)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 80°C to 84°C
Color Brown-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H4Br2O2
Linear Formula Br2C6H2(OH)CHO
Quantity 1 g
Beilstein 08,55
Merck Index 15,3032
Molecular Weight (g/mol) 279.91
Formula Weight 279.91
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Dibromosalicylaldehyde, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dibromosalicylaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.