Learn More
3,5-Dibromopyridine, 98+%
Description
3-Acetylamino-5-ethoxypyridine was prepared by converting 3, 5-dibromopyridine with sodium ethylate into 3-bromo-5-ethoxypyridine, allowing this substance to interact with ammonia and acetylating the aminoethoxypyridine thus formed with acetic anhydride. Lithiation of 3, 5-dibromopyridine with LDA and subsequent reaction with electrophiles provided 4-alkyl-3, 5-dibromopyridines 2 in high yield. Ligand was prepared by a Pd(0)-catalyzed cross-coupling reaction of 3,5-dibromopyridine and 5-tributylstannyl-3,3?-bipyridine. The synthesis of 3, 5-bis (2-indolyl) pyridine and 3-[(2-indolyl)-5-phenyl] pyridine derivatives as includes Stille or Suzuki type reactions, which were realized on the 3,5-dibromopyridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Acetylamino-5-ethoxypyridine was prepared by converting 3, 5-dibromopyridine with sodium ethylate into 3-bromo-5-ethoxypyridine, allowing this substance to interact with ammonia and acetylating the aminoethoxypyridine thus formed with acetic anhydride. Lithiation of 3, 5-dibromopyridine with LDA and subsequent reaction with electrophiles provided 4-alkyl-3, 5-dibromopyridines 2 in high yield. Ligand was prepared by a Pd(0)-catalyzed cross-coupling reaction of 3,5-dibromopyridine and 5-tributylstannyl-3,3′-bipyridine. The synthesis of 3, 5-bis (2-indolyl) pyridine and 3-[(2-indolyl)-5-phenyl] pyridine derivatives as includes Stille or Suzuki type reactions, which were realized on the 3,5-dibromopyridine.
Solubility
Soluble in Chloroform and Methanol. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, alkali metals, moisture.
Specifications
Specifications
| Melting Point | 110°C to 113°C |
| Quantity | 100 g |
| Beilstein | 108477 |
| Solubility Information | Soluble in Chloroform and Methanol. Insoluble in water. |
| Formula Weight | 236.9 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 3,5-Dibromopyridine |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.