missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromobenzyl bromide, 99%

Product Code. 11430673
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430673 1 g 1g
11440673 5 g 5g
11450673 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430673 Supplier Thermo Scientific Alfa Aesar Supplier No. A12726.03
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56908-88-4 | C7H5Br3 | 328.83 g/mol

3,5-Dibromobenzyl bromide, is an important raw material and intermediateintermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dibromobenzyl bromide, is an important raw material and intermediateintermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in toluene

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to water, oxidizing agents, bases, alcohols, amines, steel.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56908-88-4
Molecular Formula C7H5Br3
Molecular Weight (g/mol) 328.83
MDL Number MFCD00052415
InChI Key PWTFRUXTAFBWBW-UHFFFAOYSA-N
Synonym 3,5-dibromobenzyl bromide, 1,3-dibromo-5-bromomethyl benzene, benzene, 1,3-dibromo-5-bromomethyl, alpha,3,5-tribromotoluene, acmc-20andg, 3,5-dibromobenzylbromide, 3,5-dibromo benzyl bromide, 1,3-bis bromanyl-5-bromomethyl benzene
PubChem CID 143427
IUPAC Name 1,3-dibromo-5-(bromomethyl)benzene
SMILES BrCC1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 93°C to 95°C
Quantity 1 g
UN Number UN1759
Beilstein 3237827
Solubility Information Soluble in toluene
Formula Weight 328.84
Percent Purity 99%
Chemical Name or Material 3,5-Dibromobenzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.