missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromobenzotrifluoride, 96%, Thermo Scientific™

Product Code. 15497958
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497958 1 g 1g
15407968 5 g 5g
15417968 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15497958 Supplier Thermo Scientific Alfa Aesar Supplier No. H61424.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,5-Dibromobenzotrifluoride used as a intermediate in organic synthesis and pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 4°C. Storage: Keep container tightly closed. Store in a cool, dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 401-84-3
Molecular Formula C7H3Br2F3
Molecular Weight (g/mol) 303.90
MDL Number MFCD00526219
InChI Key OFNXSUANJLHGQN-UHFFFAOYSA-N
Synonym 1,3-dibromo-5-trifluoromethyl benzene, 3,5-dibromobenzotrifluoride, benzene, 1,3-dibromo-5-trifluoromethyl, pubchem11234, acmc-209jbn, ksc911m2p, 1,3-dibromobenzotrifluoride, 3,5-dibromo-, ,-trifluorotoluene, 3,5-dibromo-1-trifluoromethyl benzene, 1,3-dibromo-5-trifluoromethyl ;benzene
PubChem CID 2063489
IUPAC Name 1,3-dibromo-5-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(Br)=CC(Br)=C1

Specifications

Color Yellow
Quantity 1 g
Beilstein 2093912
Solubility Information Slightly soluble in water.
Formula Weight 303.9
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3,5-Dibromobenzotrifluoride
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.