missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromoanthranilic acid, 98+%, Thermo Scientific™

Product Code. 10228813 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10228813 5 g Glass bottle 5g
10712274 1 g Glass bottle 1g
10509613 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10228813 Supplier Thermo Scientific Acros Supplier No. 393160050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 609-85-8
Molecular Formula C7H5Br2NO2
Molecular Weight (g/mol) 294.93
MDL Number MFCD00017087
InChI Key WNABMWFLKQEGCP-UHFFFAOYSA-N
Synonym 3,5-dibromoanthranilic acid, 2-amino-3,5-dibromobenzoicacid, 3,5-dibromo-2-aminobenzoic acid, buttpark 89\07-50, 2-amino-3,5-dibromo-benzoic acid, benzoic acid, 2-amino-3,5-dibromo, acmc-20ahts, pubchem3720, 2-carboxy-4,6-dibromoaniline, ghl.pd_mitscher_leg0.1128
PubChem CID 219491
IUPAC Name 2-amino-3,5-dibromobenzoic acid
SMILES NC1=C(Br)C=C(Br)C=C1C(O)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 235°C to 236°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 294.93
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 3, 5-Dibromoanthranilic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dibromoanthranilic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.