missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dibromo-o-phenylenediamine monohydrochloride, 99%, Thermo Scientific™
£29.40 - £152.00
Chemical Identifiers
CAS | 75568-11-5 |
---|---|
Molecular Formula | C6H6Br2N2·HCl |
Molecular Weight (g/mol) | 302.4 |
MDL Number | MFCD00012967 |
InChI Key | QOZDVKFQMGARCC-UHFFFAOYSA-N |
Synonym | 3,5-dibromo-1,2-phenylenediamine monohydrochloride, 3,5-dibromobenzene-1,2-diamine hydrochloride, acmc-20andn, 1,2-diamino-3,5-dibromobenzene monohydrochloride, 3,5-bis bromanyl benzene-1,2-diamine hydrochloride, 3,5-dibromo-1,2-phenylenediamine hydrochloride, 1,2-benzenediamine,3,5-dibromo-, hydrochloride 1:1, 3,5-dibromo-ortho-phenylenediamine monohydrochloride, 3,5-dibromobenzene-1,2-diamine-hydrogen chloride 1/1, 3,5-dibromo-1,2-phenylenediaminemonohydrochloride sensitivereagentforthedeterminationofsebygc-ecd |
PubChem CID | 2724285 |
IUPAC Name | 3,5-dibromobenzene-1,2-diamine;hydrochloride |
SMILES | C1=C(C=C(C(=C1Br)N)N)Br.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10348360
|
Thermo Scientific Acros
223731000 |
100 mg | Glass bottle | ||||||
10677382
|
Thermo Scientific Acros
223735000 |
500 mg | Glass bottle |
£152.00
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
75568-11-5 | |
302.4 | |
QOZDVKFQMGARCC-UHFFFAOYSA-N | |
2724285 | |
C1=C(C=C(C(=C1Br)N)N)Br.Cl |
C6H6Br2N2·HCl | |
MFCD00012967 | |
3,5-dibromo-1,2-phenylenediamine monohydrochloride, 3,5-dibromobenzene-1,2-diamine hydrochloride, acmc-20andn, 1,2-diamino-3,5-dibromobenzene monohydrochloride, 3,5-bis bromanyl benzene-1,2-diamine hydrochloride, 3,5-dibromo-1,2-phenylenediamine hydrochloride, 1,2-benzenediamine,3,5-dibromo-, hydrochloride 1:1, 3,5-dibromo-ortho-phenylenediamine monohydrochloride, 3,5-dibromobenzene-1,2-diamine-hydrogen chloride 1/1, 3,5-dibromo-1,2-phenylenediaminemonohydrochloride sensitivereagentforthedeterminationofsebygc-ecd | |
3,5-dibromobenzene-1,2-diamine;hydrochloride |
Specifications
75568-11-5 | |
100.0 | |
Beige | |
99% | |
Br2C6H2(NH2)2·HCl | |
13, 29 | |
QOZDVKFQMGARCC-UHFFFAOYSA-N | |
3,5-dibromobenzene-1,2-diamine;hydrochloride | |
2724285 | |
99% | |
3, 5-Dibromo-o-phenylenediamine monohydrochloride, 99% |
98.5 | |
238°C | |
Authentic | |
C6H6Br2N2·HCl | |
MFCD00012967 | |
3,5-dibromo-1,2-phenylenediamine monohydrochloride, 3,5-dibromobenzene-1,2-diamine hydrochloride, acmc-20andn, 1,2-diamino-3,5-dibromobenzene monohydrochloride, 3,5-bis bromanyl benzene-1,2-diamine hydrochloride, 3,5-dibromo-1,2-phenylenediamine hydrochloride, 1,2-benzenediamine,3,5-dibromo-, hydrochloride 1:1, 3,5-dibromo-ortho-phenylenediamine monohydrochloride, 3,5-dibromobenzene-1,2-diamine-hydrogen chloride 1/1, 3,5-dibromo-1,2-phenylenediaminemonohydrochloride sensitivereagentforthedeterminationofsebygc-ecd | |
C1=C(C=C(C(=C1Br)N)N)Br.Cl | |
302.4 | |
302.4 | |
Powder |
Safety and Handling
missing translation for 'tsca' : TSCA
RUO – Research Use Only