missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-4-methylphenol, 97%, Thermo Scientific Chemicals

Product Code. 11340154
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13979-81-2
Molecular Formula C7H6Br2O
Molecular Weight (g/mol) 265.932
MDL Number MFCD00013968
InChI Key AXCQKKVGMZCWPC-UHFFFAOYSA-N
PubChem CID 55457
IUPAC Name 3,5-dibromo-4-methylphenol
SMILES CC1=C(C=C(C=C1Br)O)Br

Specifications

Melting Point 106°C to 110°C
Quantity 1 g
Beilstein 2249692
Synonym 3,5-dibromo-p-cresol, phenol, 3,5-dibromo-4-methyl, 3,5-dibromo-4-methyl-phenol, phenol, dibromomethyl, acmc-20abwf, 3,5-dibromo-p-methylphenol, 3,5-dibromo-4-methyl phenol, phenol,3,5-dibromo-4-methyl, 3,5-bis bromanyl-4-methyl-phenol
Formula Weight 265.94
Percent Purity 97%
Chemical Name or Material 3,5-Dibromo-4-methylphenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'