missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-4-hydroxybenzoic acid, 98%

Product Code. 11463927
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463927 25 g 25g
11473927 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463927 Supplier Thermo Scientific Alfa Aesar Supplier No. A18361.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3337-62-0 | C7H4Br2O3 | 295.914 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3337-62-0
Molecular Formula C7H4Br2O3
Molecular Weight (g/mol) 295.914
MDL Number MFCD00002548
InChI Key PHWAJJWKNLWZGJ-UHFFFAOYSA-N
Synonym bromoxynylbenzoic acid, dbha, benzoic acid, 3,5-dibromo-4-hydroxy, dibrhbz, unii-95z5a9epei, 3,5-dibromo-4-hydroxybenzoate, 3,5-dibromo-4-hydroxy-benzoic acid, 3',5'-dibromo-4-hydroxybenzoic acid, 95z5a9epei, rarechem al bo 0009
PubChem CID 76857
ChEBI CHEBI:1395
IUPAC Name 3,5-dibromo-4-hydroxybenzoic acid
SMILES C1=C(C=C(C(=C1Br)O)Br)C(=O)O

Specifications

Melting Point 269°C to 273°C
Quantity 25 g
Beilstein 2416147
Formula Weight 295.93
Percent Purity 98%
Chemical Name or Material 3,5-Dibromo-4-hydroxybenzoic acid
compliance-icons
Product Identifier
  • 3,5-Dibromo-4-hydroxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3,5-Dibromo-4-hydroxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.