missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-4-hydroxybenzaldehyde, 98%, Thermo Scientific™

Catalog Number 12387093
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2973-77-5
C7H4Br2O2
279.92
MFCD00016980
SXRHGLQCOLNZPT-UHFFFAOYSA-N
benzaldehyde, 3,5-dibromo-4-hydroxy, timtec-bb sbb006529, acmc-1cr6c, 3,5-dibromo-4-hydroxy benzaldehyde, 3,5-dibromo-4-hydroxy-benzaldehyde, 4-hydroxy-3,5-dibromo benzaldehyde, 3,5-dibromo-4-hydroxybenzaldehyde, 3,5-bis bromanyl-4-oxidanyl-benzaldehyde
18100
3,5-dibromo-4-hydroxybenzaldehyde
OC1=C(Br)C=C(C=O)C=C1Br
This item is not returnable. View return policy
2973-77-5
C7H4Br2O2
279.92
MFCD00016980
SXRHGLQCOLNZPT-UHFFFAOYSA-N
benzaldehyde, 3,5-dibromo-4-hydroxy, timtec-bb sbb006529, acmc-1cr6c, 3,5-dibromo-4-hydroxy benzaldehyde, 3,5-dibromo-4-hydroxy-benzaldehyde, 4-hydroxy-3,5-dibromo benzaldehyde, 3,5-dibromo-4-hydroxybenzaldehyde, 3,5-bis bromanyl-4-oxidanyl-benzaldehyde
18100
3,5-dibromo-4-hydroxybenzaldehyde
OC1=C(Br)C=C(C=O)C=C1Br
CAS Min % 97.5
CAS Max % 100.0
Melting Point 182.0°C to 186.0°C
Color Brown to Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 279.92
Percent Purity 98%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 3, 5-Dibromo-4-hydroxybenzaldehyde, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'