missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-2-pyridinol, 98%, Thermo Scientific™

Product Code. 10692075 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10692075

Brand: Thermo Scientific Acros 379670050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 13472-81-6
MDL Number MFCD00023472
InChI Key XIFRODWVHSZAMM-UHFFFAOYSA-N
Synonym 3,5-dibromo-2-hydroxypyridine, 3,5-dibromopyridin-2-ol, 3,5-dibromo-2-pyridinol, 3,5-dibromo-2 1h-pyridinone, 2-hydroxy-3,5-dibromopyridine, 3,5-dibromo-2-pyridone, 3,5-dibromopyridine-2-ol, 2 1h-pyridinone, 3,5-dibromo, pubchem6448, acmc-209bwt
PubChem CID 642826
IUPAC Name 3,5-dibromo-1H-pyridin-2-one
SMILES C1=C(C(=O)NC=C1Br)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 204.0°C to 210.0°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H3Br2NO
Quantity 5 g
Molecular Weight (g/mol) 252.89
Formula Weight 252.89
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Dibromo-2-pyridinol
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dibromo-2-pyridinol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.