missing translation for 'onlineSavingsMsg'
Learn More

3,5-dibromo-2-hydroxybenzaldehyde, 95%, Thermo Scientific™

Product Code. 10361903 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10361903 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10361903 Supplier Thermo Scientific Supplier No. BTB02697EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 90-59-5
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C7H4Br2O2
Linear Formula Br2C6H2(OH)CHO
MDL Number MFCD00003318
Quantity 10 g
Synonym 3,5-dibromosalicylaldehyde, dalyde, 3,5-dibromosalicylaldeyde, benzaldehyde, 3,5-dibromo-2-hydroxy, 2-hydroxy-3,5-dibromobenzaldehyde, 3,5-dibromosilicylaldehyde, unii-7a260pbi3a, salicylaldehyde, 3,5-dibromo, 3,5-dibromsalicylaldehyd, 3,5-dibromo salicyclic aldehyde
InChI Key JHZOXYGFQMROFJ-UHFFFAOYSA-N
SMILES C1=C(C=C(C(=C1Br)O)C=O)Br
IUPAC Name 3,5-dibromo-2-hydroxybenzaldehyde
Molecular Weight (g/mol) 279.915
PubChem CID 7024
Formula Weight 279.91
Percent Purity 95%
Chemical Name or Material 3, 5-Dibromo-2-hydroxybenzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.