missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-1H-1,2,4-triazole, 97%, Thermo Scientific™

Product Code. 10051094
Change view
Click to view available options
Quantity:
250 mg
1 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10051094 25 g Amber Glass Bottle 25g
10041674 250 mg Amber Glass Bottle 250mg
10729595 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10051094 Supplier Thermo Scientific Maybridge Supplier No. MO07857EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7411-23-6
Molecular Formula C2HBr2N3
Molecular Weight (g/mol) 226.859
InChI Key FRAKFBWDPXYIQO-UHFFFAOYSA-N
Synonym 3,5-dibromo-4h-1,2,4-triazole, 3,5-dibromo-1,2,4-triazole, 1h-1,2,4-triazole, 3,5-dibromo, s-triazole, 3,5-dibromo, 3,5-dibromo-s-triazole, so, deblqxbskfssp@, pubchem13285, acmc-1bktu, 1h-1,4-triazole, 3,5-dibromo
PubChem CID 81904
IUPAC Name 3,5-dibromo-1H-1,2,4-triazole
SMILES C1(=NC(=NN1)Br)Br

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 226.86
Percent Purity 97%
Chemical Name or Material 3,5-Dibromo-1H-1,2,4-triazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.