missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diaminobenzoic acid dihydrochloride, 99%, Thermo Scientific™

Product Code. 10021820 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10021820 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10021820 Supplier Thermo Scientific Acros Supplier No. 151190250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 618-56-4
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C7H8N2O2·2HCl
Linear Formula (H2N)2C6H3CO2H·2HCl
MDL Number MFCD00012983
Quantity 25g
Beilstein 14, 453
Synonym 3,5-diaminobenzoic acid dihydrochloride, benzoic acid, 3,5-diamino-dihydrochloride, benzoic acid, 3,5-diamino-, hydrochloride 1:2, acmc-1b2s7, 3,5-diaminobenzoicaciddihydrochloride, 3,5-diamino benzoic acid dihydrochloride
Solubility Information Solubility in water: 0.1 g/mL (20°C).
InChI Key IGPLRBRBTUNCRT-UHFFFAOYSA-N
SMILES C1=C(C=C(C=C1N)N)C(=O)O.Cl.Cl
IUPAC Name 3,5-diaminobenzoic acid;dihydrochloride
Molecular Weight (g/mol) 225.08
PubChem CID 15556789
Formula Weight 225.08
Percent Purity 99%
Chemical Name or Material 3, 5-Diaminobenzoic acid dihydrochloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Diaminobenzoic acid dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.