missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diamino-6-chloropyrazine-2-carboxylic Acid, Mikromol™

Product Code. 30050221
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050221 25 mg 25mg
30049180 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050221 Supplier Mikromol Supplier No. MM0560.020025
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3,5-Diamino-6-chloropyrazine-2-carboxylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Diamino-6-chloropyrazine-2-carboxylic Acid
CAS 4878-36-8
Analyte or Component Names 3,5-Diamino-6-chloropyrazine-2-carboxylic Acid
Shipping Condition Room Temperature
Molecular Formula C5 H5 Cl N4 O2
Certifications/Compliance ISO 17025
Synonym Amiloride Hydrochloride Imp. B (EP), Amiloride Imp. B (EP), 3,5-Diamino-6-chloro-2-pyrazinecarboxylic Acid, Pyrazinecarboxylic acid, 3,5-diamino-6-chloro- (7CI,8CI,9CI), 3,5-Diamino-6-chloropyrazine-2-carboxylic acid, 3,5-Diamino-6-chloropyrazinecarboxylic acid, 6-Chloro-3,5-diaminopyrazine-2-carboxylic acid, Amiloride Hydrochloride Dihydrate Impurity B, Amiloride Impurity B
InChI Formula InChI=1S/C5H5ClN4O2/c6-2-4(8)10-3(7)1(9-2)5(11)12/h(H,11,12)(H4,7,8,10)
SMILES Nc1nc(N)c(nc1Cl)C(=O)O
IUPAC Name 3,5-diamino-6-chloropyrazine-2-carboxylic acid
Molecular Weight (g/mol) 188.57
Quantity 25 mg
Formula Weight 188.0101 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.