missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxyphenylacetic acid, 98%

Product Code. 11467257
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467257 5 g 5g
11457257 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11467257 Supplier Thermo Scientific Alfa Aesar Supplier No. A15015.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1611-03-6 | C16H24O3 | 264.365 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1611-03-6
Molecular Formula C16H24O3
Molecular Weight (g/mol) 264.365
MDL Number MFCD00156138
InChI Key QLMGIWHWWWXXME-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-4-hydroxyphenylacetic acid, 3,5-di-tert-butyl-4-hydroxyphenyl acetic acid, 2-3,5-di tert-butyl-4-hydroxyphenyl acetic acid, 2-3,5-di-tert-butyl-4-hydroxyphenyl acetic acid, benzeneacetic acid,3,5-bis 1,1-dimethylethyl-4-hydroxy, acmc-1bvft, 4-hydroxy-3,5-di-t-butylphenylacetic acid, 3,5-di-t-butyl-4-hydroxyphenyl-acetic acid, 4-hydroxy-3,5-di-t-butyl-phenylacetic acid, 3,5-di-tert-butyl-4-hydroxybenzeneacetic acid
PubChem CID 2775110
IUPAC Name 2-(3,5-ditert-butyl-4-hydroxyphenyl)acetic acid
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CC(=O)O

Specifications

Melting Point 154°C to 158°C
Quantity 5 g
Formula Weight 264.37
Percent Purity 98%
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxyphenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.