missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di(tert-butyl)-4-hydroxybenzonitrile, 97%, Thermo Scientific™

Product Code. 10031614
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 10031614

Brand: Thermo Scientific Maybridge BTB09888EB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1988-88-1
Molecular Formula C15H21NO
Molecular Weight (g/mol) 231.34
MDL Number MFCD00156137
InChI Key AKXIIOLURNATOC-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-4-hydroxybenzonitrile, 3,5-di tert-butyl-4-hydroxybenzonitrile, benzonitrile, 3,5-bis 1,1-dimethylethyl-4-hydroxy, benzonitrile, 3,5-di-tert-butyl-4-hydroxy, 4-cyano-2,6-di-tert-butylphenol, 3,5-bis tert-butyl-4-hydroxybenzenecarbonitrile, acmc-20a6ry, 2,6-bis tert-butyl-4-cyanophenol, 3,5-di-t-butyl-4-hydroxybenzonitrile, 4-hydroxy-3,5-di-t-butylbenzonitrile
PubChem CID 137274
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 25 g
IUPAC Name 3,5-di-tert-butyl-4-hydroxybenzonitrile
Formula Weight 231.34
Percent Purity 97%
Chemical Name or Material 3, 5-Di(tert-butyl)-4-hydroxybenzonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.