missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxybenzoic acid hexadecyl ester, TRC

Product Code. 30386004
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386004 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386004 Supplier Toronto Research Chemicals Supplier No. TRCD4937331G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxybenzoic Acid-Hexadecyl Ester
CAS 67845-93-6
Molecular Formula C31 H54 O3
Quantity 1 g
Synonym 3,5-Bis(1,1-dimethylethyl)-4-hydroxybenzoic acid hexadecyl ester, 3,5-Di-tert-butyl-4-hydroxybenzoic acid hexadecyl ester, Antioxidant 2908, Cyagard UV 2908, Cyasorb 2908, Cyasorb UV 2908, Hexadecyl 3,5-bis-tert-butyl-4-hydroxybenzoate, Hexadecyl 3,5-di-tert-butyl-4-hydroxybenzoate, Hexadecyl 4-hydroxy-3,5-di(tert-butyl)benzoate, Kemisorb 114, Light Stabilizer 2908, Palmityl 3,5-di-tert-butyl-4-hydroxybenzoate, Sabostab UV 37, Songsorb 2908PW, UV 2908, 3,5-Di-tert-butyl-4-hydroxybenzoic acid-hexadecyl ester, 3,5-Di-tert-butyl-4-hydroxybenzoic Acid-Hexadecyl Ester
InChI Formula InChI=1S/C31H54O3/c1-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-34-29(33)25-23-26(30(2,3)4)28(32)27(24-25)31(5,6)7/h23-24,32H,8-22H2,1-7H3
SMILES CCCCCCCCCCCCCCCCOC(=O)c1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C
IUPAC Name hexadecyl 3,5-ditert-butyl-4-hydroxybenzoate
Molecular Weight (g/mol) 474.76
Formula Weight 474.4073
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.