missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxybenzamide, TRC

Product Code. 30374302
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374302 500 mg
30388438 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30374302 Supplier Toronto Research Chemicals Supplier No. TRCD493690500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60632-18-0
Molecular Formula C15H23NO2
Molecular Weight (g/mol) 249.35
Synonym 3,5-Bis(1,1-dimethylethyl)-4-hydroxybenzamide
SMILES CC(C)(C)c1cc(cc(c1O)C(C)(C)C)C(=O)N

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C15H23NO2/c1-14(2,3)10-7-9(13(16)18)8-11(12(10)17)15(4,5)6/h7-8,17H,1-6H3,(H2,16,18)
Formula Weight 249.1729
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxybenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.