missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-2-hydroxybenzaldehyde, 99%

Product Code. 11450308
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450308 25 g 25g
11440308 5 g 5g
11460308 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450308 Supplier Thermo Scientific Alfa Aesar Supplier No. B23834.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 37942-07-7 | C15H22O2 | 234.339 g/mol

3,5-Di-tert-butyl-2-hydroxybenzaldehyde is used in the synthesis of Mn(III)-salen complex and its diamino precursor 5,6-diamino-5,6-dideoxy-1,2-O-isopropylidene-3-O-methyl-β-L-idofuranose, chiral Schiff base ligand for an enantioselective copper-catalyzed addition of phenyl acetylene to imines, chiral oxazolidine ligand for the enantioselective addition of diethyl zinc to aldehydes and tin Schiff base complexes with histidine analogues. It has antibacterial activity and is used in the preparation nickel complexes. It is structurally related to 3,5-di-t-butylcatechol (DTCAT) but is not as potent an activator of rat skeletal muscle ryanodine receptor Ca2+ channel (RyRC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Di-tert-butyl-2-hydroxybenzaldehyde is used in the synthesis of Mn(III)-salen complex and its diamino precursor 5,6-diamino-5,6-dideoxy-1,2-O-isopropylidene-3-O-methyl-β-L-idofuranose, chiral Schiff base ligand for an enantioselective copper-catalyzed addition of phenyl acetylene to imines, chiral oxazolidine ligand for the enantioselective addition of diethyl zinc to aldehydes and tin Schiff base complexes with histidine analogues. It has antibacterial activity and is used in the preparation nickel complexes. It is structurally related to 3,5-di-t-butylcatechol (DTCAT) but is not as potent an activator of rat skeletal muscle ryanodine receptor Ca2+ channel (RyRC).

Solubility
Soluble in DMSO and methanol.

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37942-07-7
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00191998
InChI Key RRIQVLZDOZPJTH-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-2-hydroxybenzaldehyde, 3,5-di-tert-butylsalicylaldehyde, 3,5-bis 1,1-dimethylethyl-2-hydroxy-benzaldehyde, benzaldehyde, 3,5-bis 1,1-dimethylethyl-2-hydroxy, 3,5-di-t-butylsalicylaldehyde, 3,5-di-tert-butyl-2-hydroxy-benzaldehyde, benzaldehyde, 2-hydroxy, 3,5-di t-butyl, 3,5-bis tert-butyl-2-hydroxybenzaldehyde, pubchem11247, acmc-1aete
PubChem CID 688023
IUPAC Name 3,5-ditert-butyl-2-hydroxybenzaldehyde
SMILES CC(C)(C)C1=CC(=C(C(=C1)C=O)O)C(C)(C)C

Specifications

Melting Point 59°C to 61°C
Assay Percent Range 0.99
Quantity 25 g
Beilstein 1877810
Sensitivity Air Sensitive
Solubility Information Soluble in DMSO and methanol.
Formula Weight 234.34
Percent Purity 99%
Chemical Name or Material 3, 5-Di-tert-butyl-2-hydroxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.