missing translation for 'onlineSavingsMsg'
Learn More

3-(5-Bromo-pyridin-3-yl)-[1,2,4]oxadiazole-5-carboxylic acid ethyl ester, ≥97%, Thermo Scientific™

Product Code. 10166812
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10166812 1 g Amber Glass Bottle 1g
10299872 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10166812 Supplier Thermo Scientific Maybridge Supplier No. MO07248DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 850375-34-7
Molecular Formula C10H8BrN3O3
Molecular Weight (g/mol) 298.096
MDL Number MFCD06797495
InChI Key LSHJXSGSPMZMFQ-UHFFFAOYSA-N
Synonym ethyl 3-5-bromopyridin-3-yl-1,2,4-oxadiazole-5-carboxylate, 3-5-bromo-pyridin-3-yl-1,2,4 oxadiazole-5-carboxylic acid ethyl ester, 1,2,4-oxadiazole-5-carboxylicacid, 3-5-bromo-3-pyridinyl-, ethyl ester, 3-5-bromopyridin-3-yl-1,2,4 oxadiazole-5-carboxylic acid ethyl ester
PubChem CID 7170018
IUPAC Name ethyl 3-(5-bromopyridin-3-yl)-1,2,4-oxadiazole-5-carboxylate
SMILES CCOC(=O)C1=NC(=NO1)C2=CC(=CN=C2)Br

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 298.1
Percent Purity 95%
Chemical Name or Material 3-(5-Bromo-pyridin-3-yl)-[1, 2, 4]oxadiazole-5-carboxylic acid ethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.