missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 15468255
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15468255 5 g 5g
15478255 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15468255 Supplier Thermo Scientific Alfa Aesar Supplier No. H32975.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502496-23-3
Molecular Formula C8H7ClF6N2
Molecular Weight (g/mol) 280.598
MDL Number MFCD03094165
InChI Key OPIOKSFIERNABH-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenylhydrazine hydrochloride, 3,5-ditrifluoromethylphenylhydrazine hydrochloride, 3,5-bis trifluoromethyl phenyl hydrazine hydrochloride, 3,5-bis trifluoromethyl phenylhydrazinehydrochloride, 3,5-bis trifluoromethy phenylhydrazine hcl, 3,5-bis trifluoromethyl phenylhydrazine hcl, ksc491e7p, 3,5-bis trifluoromethyl phenylhydrazine hydrochlor
PubChem CID 12494556
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]hydrazine;hydrochloride
SMILES C1=C(C=C(C=C1C(F)(F)F)NN)C(F)(F)F.Cl

Specifications

Melting Point ∽208°C (decomposition)
Quantity 5 g
Formula Weight 280.6
Percent Purity 98%
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.