missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenylhydrazine, 97%, Thermo Scientific™

Product Code. 11447177
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447177 5 g 5g
11457177 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11447177 Supplier Thermo Scientific Alfa Aesar Supplier No. A14971.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,5-Bis(trifluoromethyl)phenylhydrazine is used to measure formaldehyde concentration.

Solubility
Sparingly Soluble is water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886-35-1
Molecular Formula C8H6F6N2
Molecular Weight (g/mol) 244.14
MDL Number MFCD00051606
InChI Key OULWACKZBGBRNT-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenylhydrazine, 3,5-bis-trifluoromethyl phenylhydrazine, 3,5-bis trifluoromethyl phenyl hydrazine, 3,5-di trifluoromethyl phenylhydrazine, 1-3,5-bis trifluoromethyl phenyl hydrazine, hydrazine, 3,5-bis trifluoromethyl phenyl, pubchem1080, pubchem3289, timtec-bb sbb028449
PubChem CID 522901
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]hydrazine
SMILES NNC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 80°C to 83°C
Quantity 5 g
Beilstein 1843213
Solubility Information Sparingly Soluble is water.
Formula Weight 244.14
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenylhydrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.