missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenylacetonitrile, 97%

Product Code. 11474137
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474137 5 g 5g
11464137 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474137 Supplier Thermo Scientific Alfa Aesar Supplier No. A19576.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 85068-32-2 | C10H5F6N | 253.15 g/mol

3,5-Bis(trifluoromethyl)phenylacetonitrile may be used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Bis(trifluoromethyl)phenylacetonitrile may be used in chemical synthesis.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85068-32-2
Molecular Formula C10H5F6N
Molecular Weight (g/mol) 253.15
MDL Number MFCD00009904
InChI Key YXGWYBUKRTYHJM-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenylacetonitrile, 2-3,5-bis trifluoromethyl phenyl acetonitrile, 3,5-bis trifluoromethyl benzyl cyanide, 3,5-di trifluoromethyl benzyl cyanide, 3,5-bis trifluoromethyl-phenylacetonitrile, 3,5-bis trifluoromethyl phenyl acetonitrile, benzeneacetonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzylcyanide
PubChem CID 603657
IUPAC Name 2-[3,5-bis(trifluoromethyl)phenyl]acetonitrile
SMILES FC(F)(F)C1=CC(=CC(CC#N)=C1)C(F)(F)F

Specifications

Density 1.42
Flash Point >110°C (230°F)
Refractive Index 1.4234
Quantity 5 g
UN Number UN3276
Beilstein 6810362
Solubility Information Difficult to mix.
Formula Weight 253.15
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.