missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenol, 97%

Product Code. 11469482
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11469482

Brand: Thermo Scientific Alfa Aesar A10183.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 349-58-6 | C8H4F6O | 230.109 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349-58-6
Molecular Formula C8H4F6O
Molecular Weight (g/mol) 230.109
MDL Number MFCD00000386
InChI Key ODSXJQYJADZFJX-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenol, phenol, 3,5-bis trifluoromethyl, 3,5-di trifluoromethyl phenol, 3,5-bis-trifluoromethylphenol, 3,5-ditrifluoromethylphenol, 3,5-bis-trifluoromethyl phenol, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylenol, pubchem12483, mbt-oh, acmc-1clsr
PubChem CID 67680
IUPAC Name 3,5-bis(trifluoromethyl)phenol
SMILES C1=C(C=C(C=C1C(F)(F)F)O)C(F)(F)F

Specifications

Melting Point 20°C to 21°C
Density 1.511
Boiling Point 97°C to 98°C (50 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.415
Quantity 5 g
Beilstein 2123670
Formula Weight 230.11
Percent Purity 97%
Chemical Name or Material 3,5-Bis (trifluoromethyl)phenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.