missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenol, 97%, ACROS Organics™

Product Code. 11365336 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11365336 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11365336 Supplier Thermo Scientific Acros Supplier No. 297380025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 349-58-6
CAS Min % 96.0
CAS Max % 100.0
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H4F6O
Refractive Index 1.4140 to 1.4160 (20°C, 589nm)
MDL Number MFCD00000386
Quantity 2.5 g
Synonym 3,5-bis trifluoromethyl phenol, phenol, 3,5-bis trifluoromethyl, 3,5-di trifluoromethyl phenol, 3,5-bis-trifluoromethylphenol, 3,5-ditrifluoromethylphenol, 3,5-bis-trifluoromethyl phenol, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylenol, pubchem12483, mbt-oh, acmc-1clsr
InChI Key ODSXJQYJADZFJX-UHFFFAOYSA-N
SMILES C1=C(C=C(C=C1C(F)(F)F)O)C(F)(F)F
IUPAC Name 3,5-bis(trifluoromethyl)phenol
Molecular Weight (g/mol) 230.109
PubChem CID 67680
Formula Weight 230.11
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)phenol, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.