missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Bis(trifluoromethyl)benzyl Bromide, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | 3,5-Bis(trifluoromethyl)benzyl Bromide |
| CAS | 32247-96-4 |
| Molecular Formula | C9H5BrF6 |
| Quantity | 500 mg |
| Synonym | alpha''-Bromo-alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-mesitylene, 1-(Bromomethyl)-3,5-bis(trifluoromethyl)benzene, 1-Bromomethyl-3,5-ditrifluoromethylbenzene, 3,5-(Trifluoromethyl)benzyl Bromide, 3,5-Di(trifluoromethyl)benzyl Bromide |
| InChI Formula | InChI=1S/C9H5BrF6/c10-4-5-1-6(8(11,12)13)3-7(2-5)9(14,15)16/h1-3H,4H2 |
| SMILES | FC(F)(F)c1cc(CBr)cc(c1)C(F)(F)F |
| IUPAC Name | 1-(bromomethyl)-3,5-bis(trifluoromethyl)benzene |
| Molecular Weight (g/mol) | 307.03 |
| Formula Weight | 305.9479 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction