missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzyl bromide, 98%, Thermo Scientific™

Product Code. 11424147
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424147 25 g 25g
11414147 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424147 Supplier Thermo Scientific Alfa Aesar Supplier No. A19590.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,5-Bis(trifluoromethyl)benzyl bromide is used as derivatization reagent in detection of uracil in DNA by GC and negative chemical ionization mass spectrometry in enantioselective synthesis of non-peptidic neurokinin NK1 receptor antagonist.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32247-96-4
Molecular Formula C9H5BrF6
Molecular Weight (g/mol) 307.03
MDL Number MFCD00009905
InChI Key ATLQGZVLWOURFU-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzyl bromide, 1-bromomethyl-3,5-bis trifluoromethyl benzene, 3,5-di trifluoromethyl benzyl bromide, 3,5-bis trifluoromethyl benzylbromide, 1-bromomethyl-3,5-di trifluoromethyl benzene, 3,5-bis-trifluoromethyl benzyl bromide, benzene, 1-bromomethyl-3,5-bis trifluoromethyl, 1,3-bis trifluoromethyl-5-bromomethyl benzene
PubChem CID 122573
IUPAC Name 1-(bromomethyl)-3,5-bis(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(=CC(CBr)=C1)C(F)(F)F

Specifications

Density 1.675
Boiling Point 136°C to 140°C (14 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.445
Quantity 25 g
UN Number UN3265
Beilstein 656506
Solubility Information Difficult to mix.
Formula Weight 307.04
Percent Purity 98%
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzyl bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.