missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzyl bromide, 97%

Product Code. 10259503 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10259503 5 g Glass bottle 5g
10023590 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10259503 Supplier Thermo Scientific Acros Supplier No. 303640050
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 32247-96-4 | C9H5BrF6 | 307.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32247-96-4
Molecular Formula C9H5BrF6
Molecular Weight (g/mol) 307.03
MDL Number MFCD00009905
InChI Key ATLQGZVLWOURFU-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzyl bromide, 1-bromomethyl-3,5-bis trifluoromethyl benzene, 3,5-di trifluoromethyl benzyl bromide, 3,5-bis trifluoromethyl benzylbromide, 1-bromomethyl-3,5-di trifluoromethyl benzene, 3,5-bis-trifluoromethyl benzyl bromide, benzene, 1-bromomethyl-3,5-bis trifluoromethyl, 1,3-bis trifluoromethyl-5-bromomethyl benzene
PubChem CID 122573
IUPAC Name 1-(bromomethyl)-3,5-bis(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(=CC(CBr)=C1)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.6700g/mL
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CF3)2C6H3CH2Br
Refractive Index 1.4440 to 1.446
Quantity 5 g
Specific Gravity 1.67
Formula Weight 307.04
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)benzyl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.