missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Bis(trifluoromethyl)benzyl alcohol, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 3,5-Bis(trifluoromethyl)benzyl alcohol |
| CAS | 32707-89-4 |
| Molecular Formula | C9H6F6O |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C9H6F6O/c10-8(11,12)6-1-5(4-16)2-7(3-6)9(13,14)15/h1-3,16H,4H2 |
| SMILES | C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CO |
| IUPAC Name | (3,5-bis(trifluoromethyl)phenyl)methanol |
| Molecular Weight (g/mol) | 244.13 |
| Formula Weight | 244.0323 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction