missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzoyl chloride, 97%

Product Code. 10646101 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10646101 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10646101 Supplier Thermo Scientific Acros Supplier No. 303530050
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 785-56-8 | C9H3ClF6O | 276.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 785-56-8
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5200g/mL
Boiling Point 65°C to 67°C (12.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C9H3ClF6O
Linear Formula (CF3)2C6H3COCl
Refractive Index 1.4322 to 1.4342
MDL Number MFCD00000387
Quantity 5 g
Specific Gravity 1.52
Synonym 3,5-bis trifluoromethyl benzoyl chloride, 3,5-bis-trifluoromethylbenzoyl chloride, 3,5-di trifluoromethyl benzoyl chloride, 3,5-bis trifluoromethyl benzoylchloride, benzoyl chloride, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl benzoyl chloride, benzoylchloride, 3,5-bis trifluoromethyl, fxffr cvg exfff, 3,5-bis trifluoromethyl benzoychloride
InChI Key WAKMMQSMEDJRRI-UHFFFAOYSA-N
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)Cl
IUPAC Name 3,5-bis(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol) 276.57
PubChem CID 101856
Formula Weight 276.57
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)benzoyl chloride, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.