missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™

Product Code. 11490233
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11490233

Brand: Thermo Scientific Alfa Aesar A11401.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 785-56-8
Molecular Formula C9H3ClF6O
Molecular Weight (g/mol) 276.562
MDL Number MFCD00000387
InChI Key WAKMMQSMEDJRRI-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzoyl chloride, 3,5-bis-trifluoromethylbenzoyl chloride, 3,5-di trifluoromethyl benzoyl chloride, 3,5-bis trifluoromethyl benzoylchloride, benzoyl chloride, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl benzoyl chloride, benzoylchloride, 3,5-bis trifluoromethyl, fxffr cvg exfff, 3,5-bis trifluoromethyl benzoychloride
PubChem CID 101856
IUPAC Name 3,5-bis(trifluoromethyl)benzoyl chloride
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)Cl

Specifications

Density 1.526
Boiling Point 65°C to 67°C (12 mmHg)
Flash Point 72°C (161°F)
Refractive Index 1.4332
Quantity 25 g
UN Number UN3265
Beilstein 2593440
Sensitivity Moisture sensitive
Formula Weight 276.57
Percent Purity 97%
Chemical Name or Material 3,5-Bis (trifluoromethyl)benzoyl chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.