missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzoic acid, 98%

Product Code. 11413734
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
11413734 100 g 100g
11493724 5 g 5g
11403734 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11413734 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer A11325.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 725-89-3 | C9H4F6O2 | 258.12 g/mol

In vitro metabolism studies on a series of 3,5-bis(trifluoromethyl)benzyl ethers have identified 3,5-bis(trifluoromethyl)benzoic acid as a significant metabolite possibly arising via oxidation of the benzylic position. The reactivity of the hydroxy- propyl units was demonstrated by the esterification of por- phyrazine derivatives with 3,5-bis(trifluoromethyl)benzoic acid in the presence of dicyclohexylcarbodiimide and toluene- p-sulfonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In vitro metabolism studies on a series of 3,5-bis(trifluoromethyl)benzyl ethers have identified 3,5-bis(trifluoromethyl)benzoic acid as a significant metabolite possibly arising via oxidation of the benzylic position. The reactivity of the hydroxy- propyl units was demonstrated by the esterification of por- phyrazine derivatives with 3,5-bis(trifluoromethyl)benzoic acid in the presence of dicyclohexylcarbodiimide and toluene- p-sulfonic acid.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 725-89-3
Molecular Formula C9H4F6O2
Molecular Weight (g/mol) 258.12
MDL Number MFCD00000388
InChI Key HVFQJWGYVXKLTE-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzoic acid, 3,5-di trifluoromethyl benzoic acid, benzoic acid, 3,5-bis trifluoromethyl, bis 3,5-trifluoromethyl benzoic acid, mbt-boa, 3,5-bis-trifluoromethyl-benzoic acid, 3,5-bis-trifluoromethyl benzoic acid, 3.5-bis trifluoromethyl benzoic acid, pubchem2725
PubChem CID 12889
IUPAC Name 3,5-bis(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifikationer

Melting Point 140°C to 144°C
Density 1.42
Quantity 100 g
Beilstein 2058600
Solubility Information Slightly soluble in water.
Formula Weight 258.12
Percent Purity 98%
Chemical Name or Material 3,5-Bis (trifluoromethyl)benzoic acid

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.