missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog Number 11493724
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
725-89-3
C9H4F6O2
258.12
MFCD00000388
HVFQJWGYVXKLTE-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzoic acid, 3,5-di trifluoromethyl benzoic acid, benzoic acid, 3,5-bis trifluoromethyl, bis 3,5-trifluoromethyl benzoic acid, mbt-boa, 3,5-bis-trifluoromethyl-benzoic acid, 3,5-bis-trifluoromethyl benzoic acid, 3.5-bis trifluoromethyl benzoic acid, pubchem2725
12889
3,5-bis(trifluoromethyl)benzoic acid
OC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
This item is not returnable. View return policy
725-89-3
C9H4F6O2
258.12
MFCD00000388
HVFQJWGYVXKLTE-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzoic acid, 3,5-di trifluoromethyl benzoic acid, benzoic acid, 3,5-bis trifluoromethyl, bis 3,5-trifluoromethyl benzoic acid, mbt-boa, 3,5-bis-trifluoromethyl-benzoic acid, 3,5-bis-trifluoromethyl benzoic acid, 3.5-bis trifluoromethyl benzoic acid, pubchem2725
12889
3,5-bis(trifluoromethyl)benzoic acid
OC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

In vitro metabolism studies on a series of 3,5-bis(trifluoromethyl)benzyl ethers have identified 3,5-bis(trifluoromethyl)benzoic acid as a significant metabolite possibly arising via oxidation of the benzylic position. The reactivity of the hydroxy- propyl units was demonstrated by the esterification of por- phyrazine derivatives with 3,5-bis(trifluoromethyl)benzoic acid in the presence of dicyclohexylcarbodiimide and toluene- p-sulfonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In vitro metabolism studies on a series of 3,5-bis(trifluoromethyl)benzyl ethers have identified 3,5-bis(trifluoromethyl)benzoic acid as a significant metabolite possibly arising via oxidation of the benzylic position. The reactivity of the hydroxy- propyl units was demonstrated by the esterification of por- phyrazine derivatives with 3,5-bis(trifluoromethyl)benzoic acid in the presence of dicyclohexylcarbodiimide and toluene- p-sulfonic acid.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 140°C to 144°C
Density 1.42
Quantity 5 g
Beilstein 2058600
Solubility Information Slightly soluble in water.
Formula Weight 258.12
Percent Purity 98%
Chemical Name or Material 3,5-Bis (trifluoromethyl)benzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'