missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals

Catalog Number 10275911
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
73852-19-4
C8H5BF6O2
257.93
MFCD00051850
BPTABBGLHGBJQR-UHFFFAOYSA-N
3,5-bis trifluoromethyl phenylboronic acid, 3,5-bis trifluoromethyl benzeneboronic acid, 3,5-bis trifluoromethyl phenyl boronic acid, 3,5-bis-trifluoromethylphenylboronic acid, 3,5-bis trifluoromethylphenyl boronic acid, 3,5-bis trifluoromethyl phenylboroic acid, btfpba, 3,5-bis trifluoromethyl phenyl boranediol
156265
[3,5-bis(trifluoromethyl)phenyl]boronic acid
OB(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
This item is not returnable. View return policy
73852-19-4
C8H5BF6O2
257.93
MFCD00051850
BPTABBGLHGBJQR-UHFFFAOYSA-N
3,5-bis trifluoromethyl phenylboronic acid, 3,5-bis trifluoromethyl benzeneboronic acid, 3,5-bis trifluoromethyl phenyl boronic acid, 3,5-bis-trifluoromethylphenylboronic acid, 3,5-bis trifluoromethylphenyl boronic acid, 3,5-bis trifluoromethyl phenylboroic acid, btfpba, 3,5-bis trifluoromethyl phenyl boranediol
156265
[3,5-bis(trifluoromethyl)phenyl]boronic acid
OB(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 219°C to 226°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Formula Weight 257.93
Percent Purity 95%
Physical Form Crystals or Powder
Chemical Name or Material 3, 5-Bis(trifluoromethyl)benzeneboronic acid, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Bis(trifluoromethyl)benzeneboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'