missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 11423807
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423807 1 g 1g
11433807 5 g 5g
11443807 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423807 Supplier Thermo Scientific Alfa Aesar Supplier No. A18259.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,5-Bis(trifluoromethyl)benzaldehyde was used in the synthesis of a series of meso-3,5-bis(trifluoromethyl)phenyl-substituted expanded porphyrins.

Solubility
Insoluble in water. Soluble in many organic solvents.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 401-95-6
Molecular Formula C9H4F6O
Molecular Weight (g/mol) 242.12
MDL Number MFCD00010206
InChI Key LDWLIXZSDPXYDR-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzaldehyde, 3,5-di trifluoromethyl benzaldehyde, mbt-bad, benzaldehyde, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzaldehyde, 3,5-bis-trifluoromethyl benzaldehyde, pubchem4162, acmc-1ajhv, ksc490m1j
PubChem CID 136247
IUPAC Name 3,5-bis(trifluoromethyl)benzaldehyde
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C=O

Specifications

Density 1.47
Boiling Point 171°C
Flash Point 70°C (158°F)
Refractive Index 1.422
Quantity 1 g
Beilstein 1884058
Sensitivity Air Sensitive
Solubility Information Insoluble in water. Soluble in many organic solvents.
Formula Weight 242.12
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.