missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzaldehyde, 97%

Product Code. 10348853 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10348853 25 g Glass bottle 25g
10727783 1 g Glass bottle 1g
10657061 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10348853 Supplier Thermo Scientific Acros Supplier No. 307690250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 401-95-6 | C9H4F6O | 242.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 401-95-6
Molecular Formula C9H4F6O
Molecular Weight (g/mol) 242.12
MDL Number MFCD00010206
InChI Key LDWLIXZSDPXYDR-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzaldehyde, 3,5-di trifluoromethyl benzaldehyde, mbt-bad, benzaldehyde, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzaldehyde, 3,5-bis-trifluoromethyl benzaldehyde, pubchem4162, acmc-1ajhv, ksc490m1j
PubChem CID 136247
IUPAC Name 3,5-bis(trifluoromethyl)benzaldehyde
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4700g/mL
Boiling Point 37°C (1.3 mmHg)
Flash Point 69°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CF3)2C6H3CHO
Refractive Index 1.4215 to 1.4235
Quantity 25 g
Specific Gravity 1.47
Formula Weight 242.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)benzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.