missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 10727783
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 401-95-6
Molecular Formula C9H4F6O
Molecular Weight (g/mol) 242.12
MDL Number MFCD00010206
InChI Key LDWLIXZSDPXYDR-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzaldehyde, 3,5-di trifluoromethyl benzaldehyde, mbt-bad, benzaldehyde, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzaldehyde, 3,5-bis-trifluoromethyl benzaldehyde, pubchem4162, acmc-1ajhv, ksc490m1j
PubChem CID 136247
IUPAC Name 3,5-bis(trifluoromethyl)benzaldehyde
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4700g/mL
Boiling Point 37°C (1.3 mmHg)
Flash Point 69°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CF3)2C6H3CHO
Refractive Index 1.4215 to 1.4235
Quantity 1 g
Specific Gravity 1.47
Formula Weight 242.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)benzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'