missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)aniline, 98+%

Product Code. 10357220 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10357220 10 mL Glass bottle 10mL
10721081 50 mL Glass bottle 50mL
10453965 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10357220 Supplier Thermo Scientific Acros Supplier No. 185690100
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 328-74-5 | C8H5F6N | 229.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-74-5
Molecular Formula C8H5F6N
Molecular Weight (g/mol) 229.12
MDL Number MFCD00000394
InChI Key CDIDGWDGQGVCIB-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl aniline, 3,5-di trifluoromethyl aniline, benzenamine, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenamine, 3',5'-bis-trifluoromethyl aniline, 3,5-bis-trifluoromethylaniline, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylidine, 3,5-bis-trifluoromethyl-phenylamine, aniline, 3,5-bis trifluoromethyl
PubChem CID 9480
IUPAC Name 3,5-bis(trifluoromethyl)aniline
SMILES C1=C(C=C(C=C1C(F)(F)F)N)C(F)(F)F

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Brown-Yellow to Yellow
Density 1.4700g/mL
Boiling Point 85°C (15.0 mmHg)
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (CF3)2C6H3NH2
Refractive Index 1.4330 to 1.435
Quantity 10 mL
Specific Gravity 1.47
Formula Weight 229.12
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)aniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.