missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Bis(trifluoromethyl)acetophenone, 98%

Product Code. 11494123
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494123 5 g 5g
11484123 1 g 1g
11404133 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494123 Supplier Thermo Scientific Alfa Aesar Supplier No. A10341.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 30071-93-3 | C10H6F6O | 256.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30071-93-3
Molecular Formula C10H6F6O
Molecular Weight (g/mol) 256.15
MDL Number MFCD00009910
InChI Key MCYCSIKSZLARBD-UHFFFAOYSA-N
Synonym 3',5'-bis trifluoromethyl acetophenone, 3,5-bis trifluoromethyl acetophenone, 1-3,5-bis trifluoromethyl phenyl ethanone, 1-3,5-bis trifluoromethyl phenyl ethan-1-one, mbt-ac, 1-3,5-di trifluoromethyl phenyl ethan-1-one, 3,5-ditrifluoromethylacetophenone, 3',5'-bis-trifluoromethylacetophenone
PubChem CID 121616
SMILES CC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Density 1.422
Boiling Point 85°C (8 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.423
Quantity 5 g
Beilstein 2384789
IUPAC Name 1-[3,5-bis(trifluoromethyl)phenyl]ethanone
Formula Weight 256.15
Percent Purity 98%
Chemical Name or Material 3',5'-Bis (trifluoromethyl)acetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.