missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis[2-[[[2-[(diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]ethyl]-4H-1,2,4,6-thiatriazine 1,1-Dioxide Dimaleate, Mikromol™

Product Code. 30050561
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050561 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050561 Supplier Mikromol Supplier No. MM0029.050025
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3,5-Bis[2-[[[2-[(diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]ethyl]-4H-1,2,4,6-thiatriazine 1,1-Dioxide Dimaleate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Bis[2-[([2-[(diaminomethylene)-amino]-thiazol-4-yl]methyl)-sulfanyl]ethyl]-4H-1,2,4,6-thiatriazine 1,1-Dioxide Dimaleate
CAS 109467-08-5
Analyte or Component Names 3,5-Bis[2-[([2-[(diaminomethylene)-amino]-thiazol-4-yl]methyl)-sulfanyl]ethyl]-4H-1,2,4,6-thiatriazine 1,1-Dioxide Dimaleate
Shipping Condition Room Temperature
Molecular Formula C16 H23 N11 O2 S5 . 2 C4 H4 O4
Certifications/Compliance ISO 9001
Synonym Famotidine Imp. B (EP), Famotidine USP Related Compound B, Famotidine USP RC B, Guanidine, N,N''-[(1,1-dioxido-2H-1,2,4,6-thiatriazine-3,5-diyl)bis(2,1-ethanediylthiomethylene-4,2-thiazolediyl)]bis-, (2Z)-2-butenedioate (1:2) (9CI), Guanidine, N,N''-[(1,1-dioxido-2H-1,2,4,6-thiatriazine-3,5-diyl)bis(2,1-ethanediylthiomethylene-4,2-thiazolediyl)]bis-, (Z)-2-butenedioate (1:2), Guanidine, N,N''-[2H-1,2,4,6-thiatriazine-3,5-diylbis(2,1-ethanediylthiomethylene-4,2-thiazolediyl)]bis-, S,S-dioxide, (Z)-2-butenedioate (1:2), 2H-1,2,4,6-Thiatriazine, guanidine deriv., 3,5-Bis[2-[[[2-[(diaminomethylene)-amino]-thiazol-4-yl]methyl]-sulfanyl]ethyl]-4H-1,2,4,6-thiatriazine 1,1-Dioxide Dimaleate, Famotidine Imp. B (EP) as Dimaleate, Famotidine Related Compound B as Dimaleate, Famotidine Impurity B as Dimaleate
InChI Formula InChI=1S/C16H23N11O2S5.2C4H4O4/c17-13(18)24-15-21-9(7-32-15)5-30-3-1-11-23-12(27-34(28,29)26-11)2-4-31-6-10-8-33-16(22-10)25-14(19)20;2*5-3(6)1-2-4(7)8/h7-8H,1-6H2,(H,23,26,27)(H4,17,18,21,24)(H4,19,20,22,25);2*1-2H,(H,5,6)(H,7,8)/b;2*2-1-
SMILES NC(=Nc1nc(CSCCC2=NS(=O)(=O)N=C(CCSCc3csc(N=C(N)N)n3)N2)cs1)N.OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;2-[4-[2-[5-[2-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]ethyl]-1,1-dioxo-4H-1,2,4,6-thiatriazin-3-yl]ethylsulfanylmethyl]-1,3-thiazol-2-yl]guanidine
Molecular Weight (g/mol) 793.9
Quantity 25 mg
Formula Weight 793.0859 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.