missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Bis[2-[[[2-[(diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]ethyl]-4H-1,2,4,6-thiatriazine 1,1-Dioxide Dimaleate, Mikromol
Description
3,5-Bis[2-[[[2-[(diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]ethyl]-4H-1,2,4,6-thiatriazine 1,1-Dioxide Dimaleate, Mikromol
Specifications
Specifications
| Chemical Name or Material | 3,5-Bis[2-[([2-[(diaminomethylene)-amino]-thiazol-4-yl]methyl)-sulfanyl]ethyl]-4H-1,2,4,6-thiatriazine 1,1-Dioxide Dimaleate |
| CAS | 109467-08-5 |
| Analyte or Component Names | 3,5-Bis[2-[([2-[(diaminomethylene)-amino]-thiazol-4-yl]methyl)-sulfanyl]ethyl]-4H-1,2,4,6-thiatriazine 1,1-Dioxide Dimaleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H23 N11 O2 S5 . 2 C4 H4 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Famotidine Imp. B (EP), Famotidine USP Related Compound B, Famotidine USP RC B, Guanidine, N,N'''-[(1,1-dioxido-2H-1,2,4,6-thiatriazine-3,5-diyl)bis(2,1-ethanediylthiomethylene-4,2-thiazolediyl)]bis-, (Z)-2-butenedioate (1:2), Guanidine, N,N'''-[2H-1,2,4,6-thiatriazine-3,5-diylbis(2,1-ethanediylthiomethylene-4,2-thiazolediyl)]bis-, S,S-dioxide, (Z)-2-butenedioate (1:2), 2H-1,2,4,6-Thiatriazine, guanidine deriv., 3,5-Bis[2-[[[2-[(diaminomethylene)-amino]-thiazol-4-yl]methyl]-sulfanyl]ethyl]-4H-1,2,4,6-thiatriazine 1,1-Dioxide Dimaleate, Famotidine Imp. B (EP) as Dimaleate, Famotidine Related Compound B as Dimaleate, Famotidine Impurity B as Dimaleate |
| InChI Formula | InChI=1S/C16H23N11O2S5.2C4H4O4/c17-13(18)24-15-21-9(7-32-15)5-30-3-1-11-23-12(27-34(28,29)26-11)2-4-31-6-10-8-33-16(22-10)25-14(19)20;2*5-3(6)1-2-4(7)8/h7-8H,1-6H2,(H,23,26,27)(H4,17,18,21,24)(H4,19,20,22,25);2*1-2H,(H,5,6)(H,7,8)/b;2*2-1- |
| Solution Type | Neat |
| SMILES | NC(=Nc1nc(CSCCC2=NS(=O)(=O)N=C(CCSCc3csc(N=C(N)N)n3)N2)cs1)N.OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?