missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5,6-Trichlorosalicylic acid, 98%

Product Code. 11444427
Change view
Click to view available options
Quantity:
100g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444427 100g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11444427 Supplier Alfa Aesar™ Supplier No. B20455.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
3,5,6-Trichlorosalicylic acid is a chlorinated salicylic acid intermediate.

Solubility
It has limited solubility in water.

Notes
Desiccate at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 40932-60-3
Molecular Formula C7H3Cl3O3
MDL Number MFCD00075245
Quantity 100g
Beilstein 2940592
Synonym 3,5,6-trichlorosalicylic acid, benzoic acid, 2,3,5-trichloro-6-hydroxy, 3,5,6-trichloro-2-hydroxybenzoic acid, ghl.pd_mitscher_leg0.102, acmc-209jhp, ksc489o8r, chembl10381, 3,5,6-trichloro-salicylic acid, 2,3,5-trichloro-6-hydroxy-benzoic acid, #
InChI Key IIHCUZVBIMTHEB-UHFFFAOYSA-N
SMILES C1=C(C(=C(C(=C1Cl)Cl)C(=O)O)O)Cl
IUPAC Name 2,3,5-trichloro-6-hydroxybenzoic acid
Molecular Weight (g/mol) 241.448
PubChem CID 94511
Formula Weight 241.46
Percent Purity 98%
Chemical Name or Material 3,5,6-Trichlorosalicylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.