missing translation for 'onlineSavingsMsg'
Learn More

3,5,5-Trimethylhexanoic acid, 97%

Product Code. 11328076
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328076 500 mL 500mL
11318076 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328076 Supplier Thermo Scientific Alfa Aesar Supplier No. L13360.AP
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3302-10-1 | C9H18O2 | 158.24 g/mol

3,5,5-Trimethylhexanoic acid, is an important organic intermediate. It is also used as an an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5,5-Trimethylhexanoic acid, is an important organic intermediate. It is also used as an an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Miscible with the usual organic solvents.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3302-10-1
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00020507
InChI Key OILUAKBAMVLXGF-UHFFFAOYNA-N
Synonym hexanoic acid, 3,5,5-trimethyl, 3,5,5-trimethyl-hexanoic acid, 3,5,5-trimethylhexanoicacid, unii-aw943q219o, acmc-1assi, dsstox_cid_9254, 3,5-trimethylhexanoic acid, dsstox_rid_78735, dsstox_gsid_29254, 3,5,5 trimethylhexanoic acid
PubChem CID 90960
IUPAC Name 3,5,5-trimethylhexanoic acid
SMILES CC(CC(=O)O)CC(C)(C)C

Specifications

Melting Point -70°C
Density 0.91
Boiling Point 230°C to 232°C
Flash Point 125°C (257°F)
Odor Unpleasant
Refractive Index 1.429
Quantity 500 mL
Beilstein 1749506
Solubility Information Miscible with the usual organic solvents.
Formula Weight 158.24
Percent Purity 97%
Chemical Name or Material 3,5,5-Trimethylhexanoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3,5,5-Trimethylhexanoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3,5,5-Trimethylhexanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.