missing translation for 'onlineSavingsMsg'
Learn More

3,5,3',5'-Tetraiodo Thyrolactic Acid (~90%), TRC

Product Code. 30416871
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416871 25 mg 25mg
30410692 2.5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30416871 Supplier Toronto Research Chemicals Supplier No. TRCT29616025MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7069-47-8
Molecular Formula C15 H10 I4 O5
Molecular Weight (g/mol) 777.85
Synonym Benzenepropanoic acid, α-hydroxy-4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodo-, Lactic acid, 3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]- (6CI,7CI,8CI), α-Hydroxy-4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodobenzenepropanoic acid, 3,5,3',5'-Tetraiodothyrolactic acid
IUPAC Name 2-hydroxy-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid
SMILES OC(Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H10I4O5/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23/h1-2,4-5,12,20-21H,3H2,(H,22,23)
Formula Weight 777.6707
Chemical Name or Material 3,5,3',5'-Tetraiodo Thyrolactic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.