missing translation for 'onlineSavingsMsg'
Learn More

3-[(4S)-2,2-Dimethyl-1,3-dioxolan-4-yl]-propanol, 97%

Product Code. 10757212 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10757212 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10757212 Supplier Thermo Scientific Acros Supplier No. 339610010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51268-87-2 | C8H16O3 | 160.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 51268-87-2
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 90°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C8H16O3
Refractive Index 1.4405 to 1.4425
MDL Number MFCD01631673
Quantity 1 g
Specific Rotation +9° to +12° (20°C, 589nm) (c=1, CHCl3)
Synonym 3-4s-2,2-dimethyl-1,3-dioxolan-4-yl propan-1-ol, 1,3-dioxolane-4-propanol, 2,2-dimethyl-, s, s-3-2,2-dimethyl-1,3-dioxolan-4-yl propan-1-ol, 3-4s-2,2-dimethyl-1,3-dioxolan-4-yl-propanol, 1,3-dioxolane-4-propanol,2,2-dimethyl-, 4s, 3-2,2-dimethyl-1,3-dioxolane-4 s-yl-propanol, 3-2,2-dimethyl-1,3-dioxolane-4beta-yl-1-propanol
InChI Key BSLDYXOPGSOQGZ-ZETCQYMHSA-N
SMILES CC1(OCC(O1)CCCO)C
IUPAC Name 3-[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]propan-1-ol
Molecular Weight (g/mol) 160.21
PubChem CID 2733890
Formula Weight 160.21
Percent Purity 97%
Physical Form Oily Liquid
Chemical Name or Material 3-[(4S)-2, 2-Dimethyl-1, 3-dioxolan-4-yl]-propanol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-[(4S)-2,2-Dimethyl-1,3-dioxolan-4-yl]-propanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.