missing translation for 'onlineSavingsMsg'
Learn More

3-(4H-1,2,4-Triazol-4-yl)-benzoic Acid, TRC

Product Code. 30455605
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455605 100 mg -
30453426 50 mg 50mg
30450382 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455605 Supplier Toronto Research Chemicals Supplier No. TRCT899353100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335255-80-6
Molecular Formula C9H7N3O2
Molecular Weight (g/mol) 189.17
IUPAC Name 3-(1,2,4-triazol-4-yl)benzoic acid
SMILES OC(=O)c1cccc(c1)n2cnnc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H7N3O2/c13-9(14)7-2-1-3-8(4-7)12-5-10-11-6-12/h1-6H,(H,13,14)
Formula Weight 189.0538
Chemical Name or Material 3-(4H-1,2,4-Triazol-4-yl)-benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.