missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Pyridyl)propanoic acid, 95%, Thermo Scientific™

Product Code. 15389059 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15389059

Brand: Thermo Scientific Acros 457270010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6318-43-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
InChI Key WSXGQYDHJZKQQB-UHFFFAOYSA-N
Synonym 3-pyridin-4-yl propanoic acid, 3-pyridin-4-yl-propionic acid, 4-pyridinepropanoic acid, 4-pyridinepropionic acid, 3-4-pyridinyl propanoic acid, 3-4-pyridyl propanoic acid, 3-4-pyridyl propionic acid, 3-pyridin-4-yl propionic acid, 4-carboxyethylpyridine, acmc-1b6yz
PubChem CID 233243
IUPAC Name 3-pyridin-4-ylpropanoic acid
SMILES C1=CN=CC=C1CCC(=O)O

Specifications

Melting Point 229.0°C to 233.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 151.17
Percent Purity 95%
Chemical Name or Material 3-(4-Pyridyl)propanoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.