missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine, TRC

Product Code. 30413407
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413407 25 g -
30424674 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30413407 Supplier Toronto Research Chemicals Supplier No. TRCP30139025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 330786-24-8
Molecular Formula C17H13N5O
Molecular Weight (g/mol) 303.32
Synonym 1H-Pyrazolo[3,4-d]pyrimidin-4-amine, 3-(4-phenoxyphenyl)- (9CI, ACI), 3-(4-Phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine (ACI), 3-(4-Phenoxyphenyl)-1H-pyrazolo(3,4 d)pyrimidin-4-amine, 3-(4-Phenoxyphenyl)-1H-pyrazolo(3,4 d)pyrimidin-4-amine
IUPAC Name 3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
SMILES Nc1ncnc2[nH]nc(c3ccc(Oc4ccccc4)cc3)c12

Specifications

Quantity 25 g
InChI Formula InChI=1S/C17H13N5O/c18-16-14-15(21-22-17(14)20-10-19-16)11-6-8-13(9-7-11)23-12-4-2-1-3-5-12/h1-10H,(H3,18,19,20,21,22)
Formula Weight 303.112
Chemical Name or Material 3-(4-Phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.