missing translation for 'onlineSavingsMsg'
Learn More

3,4-O-Isopropylidene Clindamycin B, TRC

Product Code. 30396578
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396578 10 mg 10mg
30393123 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396578 Supplier Toronto Research Chemicals Supplier No. TRCI82487510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-O-Isopropylidene Clindamycin B
Analyte or Component Names 3,4-O-Isopropylidene Clindamycin B
Shipping Condition Room Temperature
Molecular Formula C20 H35 Cl N2 O5 S
Synonym (2S-trans)-Methyl 7-Chloro-6,7,8-trideoxy-3,4-O-(1-methylethylidene)-6-[[(1-methyl-4-ethyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-L-threo-alpha-D-galacto-octopyranoside
InChI Formula InChI=1S/C20H35ClN2O5S/c1-7-11-8-12(23(5)9-11)18(25)22-13(10(2)21)15-17-16(27-20(3,4)28-17)14(24)19(26-15)29-6/h10-17,19,24H,7-9H2,1-6H3,(H,22,25)/t10-,11-,12?,13+,14?,15+,16?,17-,19+/m0/s1
Solution Type Neat
SMILES CC[C@H]1CC(N(C)C1)C(=O)N[C@H]([C@H](C)Cl)[C@H]2O[C@H](SC)C(O)C3OC(C)(C)O[C@@H]23
IUPAC Name (4S)-N-[(1S,2S)-1-[(3aS,4R,6R)-7-hydroxy-2,2-dimethyl-6-methylsulfanyl-4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-4-yl]-2-chloropropyl]-4-ethyl-1-methylpyrrolidine-2-carboxamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 451.02
Quantity 10 mg
Formula Weight 450.196
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.