missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-O-Isopropylidene Clindamycin B, TRC
Specifications
Specifications
| Chemical Name or Material | 3,4-O-Isopropylidene Clindamycin B |
| Analyte or Component Names | 3,4-O-Isopropylidene Clindamycin B |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H35 Cl N2 O5 S |
| Synonym | (2S-trans)-Methyl 7-Chloro-6,7,8-trideoxy-3,4-O-(1-methylethylidene)-6-[[(1-methyl-4-ethyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-L-threo-alpha-D-galacto-octopyranoside |
| InChI Formula | InChI=1S/C20H35ClN2O5S/c1-7-11-8-12(23(5)9-11)18(25)22-13(10(2)21)15-17-16(27-20(3,4)28-17)14(24)19(26-15)29-6/h10-17,19,24H,7-9H2,1-6H3,(H,22,25)/t10-,11-,12?,13+,14?,15+,16?,17-,19+/m0/s1 |
| Solution Type | Neat |
| SMILES | CC[C@H]1CC(N(C)C1)C(=O)N[C@H]([C@H](C)Cl)[C@H]2O[C@H](SC)C(O)C3OC(C)(C)O[C@@H]23 |
| IUPAC Name | (4S)-N-[(1S,2S)-1-[(3aS,4R,6R)-7-hydroxy-2,2-dimethyl-6-methylsulfanyl-4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-4-yl]-2-chloropropyl]-4-ethyl-1-methylpyrrolidine-2-carboxamide |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?